4-(2-Carboxyethyl)benzeneboronic acid, 97%
4-(2-Carboxyethyl)benzeneboronic acid, 97%
4-(2-Carboxyethyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

4-(2-Carboxyethyl)benzeneboronic acid, 97%

CAS: 166316-48-9 | C9H11BO4 | 193.993 g/mol
製品番号(カタログ番号) L17485.03
または、製品番号L17485-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS166316-48-9
IUPAC Name3-[4-(dihydroxyboranyl)phenyl]propanoic acid
Molecular FormulaC9H11BO4
InChI KeyVPSARXNVXCRDIV-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(CCC(O)=O)C=C1
さらに表示
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Assay (HPLC)≥96.0%
Melting Point (clear melt)140-163°C
4-(2-Carboxyethyl)benzeneboronic acid is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Carboxyethyl)benzeneboronic acid is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only