7-Fluoroindole, 97%
7-Fluoroindole, 97%
7-Fluoroindole, 97%
Thermo Scientific Chemicals

7-Fluoroindole, 97%

CAS: 387-44-0 | C8H6FN | 135.14 g/mol
製品番号(カタログ番号) L17621.03
または、製品番号L17621-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS387-44-0
Appearance (Color)White to cream to yellow to pale brown
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or fused solid or flakes
Melting Point (clear melt)61.0-70.0?C
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in methanol. (almost transparency)

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Joseph B. Blair.; Deborah Kurrasch-Orbaugh.; Danuta Marona-Lewicka.; Medhane G. Cumbay.; Val J. Watts.; Eric L. Barker.; David E. Nichols. Effect of Ring Fluorination on the Pharmacology of Hallucinogenic Tryptamines.J. Med. Chem. 2000, 43 (24),4701-4710 .