2-Amino-5-bromo-3,4-dimethylpyridine, 97%
2-Amino-5-bromo-3,4-dimethylpyridine, 97%
2-Amino-5-bromo-3,4-dimethylpyridine, 97%
Thermo Scientific Chemicals

2-Amino-5-bromo-3,4-dimethylpyridine, 97%

CAS: 374537-97-0 | C7H9BrN2 | 201.07 g/mol
製品番号(カタログ番号) L17672.03
または、製品番号L17672-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS374537-97-0
Appearance (Color)Pale brown to brown
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)129.0-138.0?C
It is an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Evgeny Kiselev.; Keli Agama.; Yves Pommier.; Mark Cushman. Azaindenoisoquinolines as Topoisomerase I Inhibitors and Potential Anticancer Agents: A Systematic Study of Structure-Activity Relationships.J. Med. Chem. 2012, 55 (4),1682-1697 .