6-Bromoindole, 98%, Thermo Scientific Chemicals
6-Bromoindole, 98%, Thermo Scientific Chemicals
6-Bromoindole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromoindole, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
L17703.03
または、製品番号L17703-03
1 g
製品番号(カタログ番号) L17703.03
または、製品番号L17703-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS52415-29-9
IUPAC Name6-bromo-1H-indole
Molecular FormulaC8H6BrN
InChI KeyMAWGHOPSCKCTPA-UHFFFAOYSA-N
SMILESBrC1=CC=C2C=CNC2=C1
さらに表示
FormPowder
Melting Point (clear melt)90.0-96.0?C
Identification (FTIR)Conforms
Appearance (Color)Pale cream to cream to yellow to brown or pink
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only