2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%
2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%
Thermo Scientific Chemicals

2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%

CAS: 1195-66-0 | C7H15BO3 | 158.00 g/mol
製品番号(カタログ番号) L19056.06
または、製品番号L19056-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1195-66-0
Refractive Index1.4075-1.4125 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
It is employed in the synthesis, optical, and electrochemical properties of novel copolymers on the basis of benzothiadiazole and electron-rich arene units.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis, optical, and electrochemical properties of novel copolymers on the basis of benzothiadiazole and electron-rich arene units.

Notes
Moisture sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
RUO – Research Use Only

General References:

  1. Carles Ayats, et al. Synthesis of the pyrrolo[2,3-c]carbazole core of the dictyodendrins.Org. Biomol. Chem.,2009,7(5), 860-862.
  2. M. Jayakannan, et al. Synthesis, optical, and electrochemical properties of novel copolymers on the basis of benzothiadiazole and electron-rich arene units.J. Polym. Sci.,2002,40(14), 2360-2372.
  3. Boronation reagent; see 2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, L17278 .