2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 98%
2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 98%
2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 98%
2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 98%
Thermo Scientific Chemicals

2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 98%

CAS: 86604-75-3 | C9H13Cl2NO | 222.109 g/mol
製品番号(カタログ番号) L19453.06
または、製品番号L19453-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS86604-75-3
IUPAC Name2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride
Molecular FormulaC9H13Cl2NO
InChI KeyLCJDHJOUOJSJGS-UHFFFAOYSA-N
SMILESCl.COC1=C(C)C(CCl)=NC=C1C
さらに表示
FormPowder or crystalline powder
Appearance (Color)White to pale cream
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only