(S)-(-)-3-Aminopyrrolidine, 99%, ee 99%
(S)-(-)-3-Aminopyrrolidine, 99%, ee 99%
(S)-(-)-3-Aminopyrrolidine, 99%, ee 99%
Thermo Scientific Chemicals

(S)-(-)-3-Aminopyrrolidine, 99%, ee 99%

CAS: 128345-57-3 | C4H10N2 | 86.14 g/mol
製品番号(カタログ番号) L19500.03
または、製品番号L19500-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS128345-57-3
IUPAC Namepyrrolidin-3-amine
Molecular FormulaC4H10N2
InChI KeyNGXSWUFDCSEIOO-UHFFFAOYNA-N
SMILESNC1CCNC1
さらに表示
Enantiomeric excess≥98.5%
Optical Rotation-18° to - 22° (neat)
Refractive Index1.4880-1.4910 @ 20°C
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to yellow to brown
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only