2,3-Dibromopyridine, 97%
2,3-Dibromopyridine, 97%
2,3-Dibromopyridine, 97%
Thermo Scientific Chemicals

2,3-Dibromopyridine, 97%

CAS: 13534-89-9 | C5H3Br2N | 236.894 g/mol
製品番号(カタログ番号) L19527.06
または、製品番号L19527-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13534-89-9
IUPAC Name2,3-dibromopyridine
Molecular FormulaC5H3Br2N
InChI KeySLMHHOVQRSSRCV-UHFFFAOYSA-N
SMILESBrC1=CC=CN=C1Br
さらに表示
FormPowder
Appearance (Color)White to cream to pale brown
Assay (GC)≥96.0%
Melting Point (clear melt)55.0-64.0?C
It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Store in dry condition. Keep oxidizing agents.
RUO – Research Use Only

General References:

  1. Kristof T. J. Loones.; Bert U. W. Maes.; Caroline Meyers.; Joke Deruytter. Orthogonal and Auto-Tandem Catalysis: Synthesis of Dipyrido[1,2-a:2‘,3‘-d]imidazole and Its Benzo and Aza Analogues via Inter- and Intramolecular C-N Bond Formation.J. Org. Chem. 2006, 71 (1), 260-264 .
  2. Philippe C. Gros.; Fatima Elaachbounia. Bromine-lithium exchange under non-cryogenic conditions: TMSCH2Li-LiDMAE promoted C-2 lithiation of 2,3-dibromopyridine.Chem. Commun. 2008, (39),4813-4815 .