Thermo Scientific Chemicals

2-Phenylethyl-1-boronic acid pinacol ester, 99%, Thermo Scientific Chemicals

Catalog number: L19530.03
1 g, Each
Thermo Scientific Chemicals

2-Phenylethyl-1-boronic acid pinacol ester, 99%, Thermo Scientific Chemicals

Catalog number: L19530.03
1 g, Each
Quantity
Catalog number: L19530.03
also known as L19530-03
Price (USD)
Price: 204.00
Online price: 176.65
Your price:
Quantity
-

Chemical Identifiers

CAS
165904-22-3
IUPAC Name
4,4,5,5-tetramethyl-2-(2-phenylethyl)-1,3,2-dioxaborolane
Molecular Formula
C14H21BO2
InChI Key
LVLQNRWCBBIVHR-UHFFFAOYSA-N
SMILES
CC1(C)OB(CCC2=CC=CC=C2)OC1(C)C
Form
Fused solid or liquid as melt
Appearance (Color)
White or clear colorless
Assay (GC)
≥98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text