Isoquinoline-5-carboxaldehyde, 97%
Isoquinoline-5-carboxaldehyde, 97%
Isoquinoline-5-carboxaldehyde, 97%
Thermo Scientific Chemicals

Isoquinoline-5-carboxaldehyde, 97%

CAS: 80278-67-7 | C10H7NO | 157.172 g/mol
製品番号(カタログ番号) L19636.MD
または、製品番号L19636-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS80278-67-7
IUPAC Nameisoquinoline-5-carbaldehyde
Molecular FormulaC10H7NO
InChI KeyILRSABOCKMOFGW-UHFFFAOYSA-N
SMILESO=CC1=C2C=CN=CC2=CC=C1
さらに表示
Appearance (Color)Yellow to brown
FormPowder
Free acid (titration)≤3%
Melting Point (clear melt)113.0-122.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only