3-Amino-2-bromopyridine, 97%
3-Amino-2-bromopyridine, 97%
3-Amino-2-bromopyridine, 97%
3-Amino-2-bromopyridine, 97%
Thermo Scientific Chemicals

3-Amino-2-bromopyridine, 97%

CAS: 39856-58-1 | C5H5BrN2 | 173.013 g/mol
製品番号(カタログ番号) L19839.14
または、製品番号L19839-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS39856-58-1
IUPAC Name2-bromopyridin-3-amine
Molecular FormulaC5H5BrN2
InChI KeyHKDVVTLISGIPFE-UHFFFAOYSA-N
SMILESNC1=CC=CN=C1Br
さらに表示
Assay (GC)≥96.0%
Appearance (Color)Pale yellow to orange or cream to dark brown
FormCrystals or powder or crystalline powder
Melting Point (clear melt)74.0-80.0?C
Used in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles. pyrrole derivatives ahs been prepared using 3-Amino-2-bromopyridine. Used in P-C coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles. pyrrole derivatives ahs been prepared using 3-Amino-2-bromopyridine. Used in P-C coupling reaction.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Mohamed Shaker S. Adam,; Olaf Kühl,; Markus K. Kindermann,; Joachim W. Heinicke,; Peter G. Jones. 3-Amino- and 3-acylamido-2-phosphonopyridines: synthesis by Pd-catalyzed P-C coupling, structure and conversion to pyrido[b]-anellated Pdouble bond; length as m-dashC-N heterocycles. Tetrahedron. 2008, 64(34), 7960-7967.