2-Bromopyridine-4-carboxamide, 97%
2-Bromopyridine-4-carboxamide, 97%
2-Bromopyridine-4-carboxamide, 97%
Thermo Scientific Chemicals

2-Bromopyridine-4-carboxamide, 97%

CAS: 29840-73-1 | C6H5BrN2O | 201.023 g/mol
製品番号(カタログ番号) L19854.06
または、製品番号L19854-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS29840-73-1
IUPAC Name2-bromopyridine-4-carboxamide
Molecular FormulaC6H5BrN2O
InChI KeyKCELTDZFDHPTBI-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=NC(Br)=C1
さらに表示
Appearance (Color)White to brown
Melting Point (clear melt)183.0-193.0?C
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
2-Bromopyridine-4-carboxamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyridine-4-carboxamide is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only