2-Chloro-4-cyanopyridine, 97%
2-Chloro-4-cyanopyridine, 97%
2-Chloro-4-cyanopyridine, 97%
Thermo Scientific Chemicals

2-Chloro-4-cyanopyridine, 97%

CAS: 33252-30-1 | C6H3ClN2 | 138.554 g/mol
製品番号(カタログ番号) L19858.06
または、製品番号L19858-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS33252-30-1
IUPAC Name2-chloropyridine-4-carbonitrile
Molecular FormulaC6H3ClN2
InChI KeyQRXBTPFMCTXCRD-UHFFFAOYSA-N
SMILESClC1=CC(=CC=N1)C#N
さらに表示
Appearance (Color)White to dark cream to cream to pale brown
Assay (GC)≥96.0%
FormPowder
Melting Point (clear melt)67.0-76.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only