2-Amino-5-fluoropyridine, 97%
2-Amino-5-fluoropyridine, 97%
2-Amino-5-fluoropyridine, 97%
Thermo Scientific Chemicals

2-Amino-5-fluoropyridine, 97%

CAS: 21717-96-4 | C5H5FN2 | 112.11 g/mol
製品番号(カタログ番号) L20010.03
または、製品番号L20010-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS21717-96-4
IUPAC Name5-fluoropyridin-2-amine
Molecular FormulaC5H5FN2
InChI KeyYJTXQLYMECWULH-UHFFFAOYSA-N
SMILESNC1=CC=C(F)C=N1
さらに表示
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder
Appearance (Color)Pale cream to cream to brown or yellow
Assay (GC)≥96.0%
Melting Point (clear melt)91.5-100.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only