Employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Takashi Sasaki; Jason M. Guerrero; Ashley D. Leonard; James M. Tour. Nanotrains and self-assembled two-dimensional arrays built from carboranes linked by hydrogen bonding of dipyridones. Nano Research. 2008, 1(5), 412-419.
- Takashi Sasaki; Jason M. Guerrero; James M. Tour. The assembly line: self-assembling nanocars. Tetrahedron. 2008, 64(36), 412-419.