5-Bromopyridine-3-boronic acid, tech. 85%, Thermo Scientific Chemicals
5-Bromopyridine-3-boronic acid, tech. 85%, Thermo Scientific Chemicals
5-Bromopyridine-3-boronic acid, tech. 85%, Thermo Scientific Chemicals
5-Bromopyridine-3-boronic acid, tech. 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromopyridine-3-boronic acid, tech. 85%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
L20084.03
または、製品番号L20084-03
1 g
製品番号(カタログ番号) L20084.03
または、製品番号L20084-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS452972-09-7
FormPowder
Assay (HPLC)≥80.0%
Proton NMRConforms to structure
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥80.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only