Thermo Scientific Chemicals

6-Bromopyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: L20085.MD
250 mg, Each
Thermo Scientific Chemicals

6-Bromopyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: L20085.MD
250 mg, Each
Quantity
Catalog number: L20085.MD
also known as L20085-MD
Price (USD)
Price: 56.20
Online price: 48.65
Your price:
Quantity
-

Chemical Identifiers

CAS
223463-14-7
Proton NMR
Conforms to structure
Form
Powder
Assay (HPLC)
≥94.0%
Appearance (Color)
Cream
Assay (Aqueous acid-base Titration)
≥94.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text