1-Bromo-4-tert-butoxybenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-tert-butoxybenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-tert-butoxybenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-4-tert-butoxybenzene, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
L20158.06
または、製品番号L20158-06
5 g
製品番号(カタログ番号) L20158.06
または、製品番号L20158-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS60876-70-2
IUPAC Name1-bromo-4-(tert-butoxy)benzene
Molecular FormulaC10H13BrO
InChI KeyQIWQHUCUWNGYDZ-UHFFFAOYSA-N
SMILESCC(C)(C)OC1=CC=C(Br)C=C1
さらに表示
FormCrystalline powder or Low melting solid
Appearance (Color)White to pale yellow.
Assay (GC)≥97.5%
Melting Point (clear melt)32-37?C
It is used in the stereoselective synthesis of four stereoisomers of β-Methoxytyrosine, a component of callipeltin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the stereoselective synthesis of four stereoisomers of β-Methoxytyrosine, a component of callipeltin A.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Bryan Li, et al. Aqueous Phosphoric Acid as a Mild Reagent for Deprotection of tert-Butyl Carbamates, Esters, and Ethers.J. Org. Chem.,2006,71(24), 9045-9050.
  2. Darren B. Hansen, et al. Stereoselective Synthesis of Four Stereoisomers of β-Methoxytyrosine, a Component of Callipeltin A.J. Org. Chem.,2005,70(8), 3120-3126.