iConnect Programmable Temperature Vaporizer (PTV) Injector Module with integrated Backflush for TRACE 1600 Series GC
iConnect Programmable Temperature Vaporizer (PTV) Injector Module with integrated Backflush for TRACE 1600 Series GC
iConnect Programmable Temperature Vaporizer (PTV) Injector Module with integrated Backflush for TRACE 1600 Series GC
iConnect Programmable Temperature Vaporizer (PTV) Injector Module with integrated Backflush for TRACE 1600 Series GC
iConnect Programmable Temperature Vaporizer (PTV) Injector Module with integrated Backflush for TRACE 1600 Series GC
Thermo Scientific™
iConnect Programmable Temperature Vaporizer (PTV) Injector Module with integrated Backflush for TRACE 1600 Series GC
Manage a difficult and heavy matrix through integrated backflushing, protect from contamination, increase throughput and decrease running costs.
Have Questions?
Catalog NumberQuantity
MI-190000-0020Each
Catalog number MI-190000-0020
Price (BRL)
-
Quantity:
Each
Simplify backflush injection technique and increase the robustness of your GC system, preserving analytical column lifetime while maximizing laboratory throughput. Combine backflush capability to normal or large volume injections through pre-column, post-column, or mid-column backflush configuration to eliminate unwanted high boilers. The integrated backflush technolology of the Thermo Scientific™ iConnect™ PTV-BKF injector module removes the complexity and cost of an additional auxiliary gas module, and facilitates method setup thanks to the self-adjusting pressure during backflushing.
Save analysis time and gain in robustness against heavy contamination. Experience the simplicity of backflush injection to:
  • Extend instrument uptime and reduce maintenance by preventing column and detector contamination through removal of unwanted high boiling compounds
  • Confidently select the best backflush configuration for your sample composition, with correct and self-adjusted pneumatic settings
  • Quickly and easily install the iConnect PTV-BKF module, thanks to the modular design of the TRACE 1600 Series GC
  • Set up your columns connection with the use of the Thermo Scientific™ microfluidic device, based on SGE's SilFlow technology, easily accessible, fitting neatly within the column oven
  • Control all the injection steps through the Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) software
Specifications
QuantityEach
For Use With (Equipment)Trace 1600 Series
Injection ModeHot/Cold Split, Splitless, Large-Volume (Solvent Split) injection with reversal flow (backflush)
TypePTV Injector with integrated backflush
Unit SizeEach