TaqMan™ Reverse Transcription Reagents
Invitrogen™

TaqMan™ Reverse Transcription Reagents

TaqMan Reverse Transcription Reagents provide the necessary reagents for reverse transcriptase-PCR (RT-PCR). Random hexamers, oligo (dT)16, and reverse primers are included for cDNA synthesis flexibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
N8080234200 Reactions at 10 μL
43041342000 Reactions at 10 μL
Catalog number N8080234
Price (USD)
861.65
Online exclusive
968.00
Save 106.35
Each
No. of Reactions:
200 Reactions at 10 μL
Price (USD)
861.65
Online exclusive
968.00
Save 106.35
Each

TaqMan Reverse Transcription Reagents provide the necessary reagents for reverse transcriptase-PCR (RT-PCR). Random hexamers, oligo (dT)16, and reverse primers are included for cDNA synthesis flexibility. These reagents are designed for reverse transcription and PCR amplification of a specific target RNA from either total RNA or mRNA. The reagents in this kit facilitate two-step RT-PCR, which is performed in two separate reactions and is useful when detecting multiple transcripts from a single cDNA reaction or when storing a portion of the cDNA.

Advantages of TaqMan Reverse Transcription Reagents

  • Contains MultiScribe Reverse Transcriptase, a recombinant Moloney murine leukemia virus (MMLV) reverse transcriptase
  • Includes random hexamers and oligo (dT)16 for flexibility
  • Individual components provide versatility in assay setup
  • Compatible with two-step RT-PCR reactions

Using TaqMan Reverse Transcription Reagents

  • TaqMan Reverse Transcription Reagents provide all the necessary components for reverse transcription of RNA to cDNA
  • For two-step RT-PCR, random hexamers, oligo (dT)16, and sequence-specific reverse primers can all be used
  • The choice of primers for reverse transcription is best made after experimentally evaluating all three priming systems
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatKit
No. of Reactions200 Reactions at 10 μL
Optimal Reaction Temperature42°C
Quantity200 Reactions
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseMultiScribe
Ribonuclease H ActivityYes
Shipping ConditionDry Ice
Size (Final Product)Up to 5 kb
Starting MaterialRNA
TechniqueReverse Transcription
Concentration50 U/μL
For Use With (Application)Real Time PCR (qPCR)
Unit SizeEach
Contents & Storage

• MultiScribe Reverse Transcriptase, 100 μL (50 U/μL)
• RNase Inhibitor, 100 μL (20 U/μL)
• dNTP Mixture, 1 mL (2.5 mM each dNTP)
• Oligo (dT)16, 100 μL (50 μM)
• Random Hexamers, 100 μL (50 μM)
• 10X RT Buffer, 1.5 mL
• MgCl2 Solution, 1.5 mL

Store at –25°C to –15°C.

Frequently asked questions (FAQs)

Can the RT buffer for TaqMan Reverse Transcription Reagents kit (Cat. No. N8080234) be purchased separately?

Yes, it is the same as the GeneAmp 10X PCR Buffer II & MgCl2 (Cat. No. N8080010).

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.