Bolt™ Welcome Pack with iBlot™ 3 System - FAQs
Bolt™ Welcome Pack with iBlot™ 3 System - FAQs
View additional product information for Bolt™ Welcome Pack with iBlot™ 3 System - FAQs (NW0412AIB3)
1 product FAQs found
Do you offer additional years of service for the iBlot 3 Western Transfer Device?
No, we do not provide service contracts or extended warranties for these devices. The warranty is limited to 1 year from the date of shipment.