Product Image
Thermo Scientific™

Eutech™ PCSTestr 35 Multi-Parameter Pocket Tester

Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.
製品番号(カタログ番号)タイプ
PCSTEST35Multi-Parameter Pocket Tester
製品番号(カタログ番号) PCSTEST35
価格(JPY)
-
タイプ:
Multi-Parameter Pocket Tester
No need to change sensor every time you switch parameters – The Eutech™ PCSTestr 35 Multi-Parameter Pocket Tester measures five parameters (measures pH, Conductivity, TDS, Salinity and Temperature). For general applications: Ideal for quick and accurate pH checks in pools and spas, aquariums and hydroponics operations, or anywhere frequent pH testing is required. PCSTestr 35 meets requirements for many standard environmental and ASTM test methods. For industrial applications: Use for cooling towers, food processing water testing and process/wastewater testing in metal finishing, photo development, printing and chemical industries. For educational applications: Useful in schools, many laboratory applications and ecology studies.

Features

  • Full pH rangemeasurement at up to 2-decimal places
  • Low,medium and high Conductivity/ TDS ranges – testermeasures pure water
  • multi-range Salinitymeasurements of up to 10.00 ppt or 1.00 %
  • Up to 5-point pH calibration and 3-point Conductivity/ TDS/ Salinity calibration
  • Adjustable TDS factor (0.40 to 1.00) and Temperature Coefficient Feature (0.0 to 10.0%)
  • Temperature sensor housed within unique dome-shaped casing for faster ATC response and fastermeasurements
  • ATC/mTC option
  • Scroll-menu with four buttons control for stress-free navigation
  • Longest-lasting tester in themarket! No tools required to replace sensor.
  • 'One-press Parameter Toggle': Switching parameters is just a press of the button.
  • Certified IP67 Waterproof – Lightweight tester even floats for easy retrieval.

Specifications

  • pH Range: 0.00 to 14.00 pH
  • Resolution: 0.01 pH
  • Accuracy: ±0.01 pH
  • pH Calibration Points: Up to 5 (Auto)
  • pH Buffer Options: USA, NIST
  • Conductivity Range: .. to 199.9 μS/cm, 200 to 1999 μS/cm, 2.00 to 20.00mS/cm
  • Resolution: 0.1 μS/cm, 1 μS/cm, 0.01mS/cm
  • Accuracy: ±1% Full Scale
  • Conductivity Calibration Points: 3 Auto, 3manual
  • Normalization Temperature: 25.0 °C
  • Temperature Coefficient: 0.0 to 10.0%/°C
  • Auto-Ranging: Yes
  • TDS Range: 0.0 to 99.9 ppm, 100 to 999 ppm, 1.00 to 10.00 ppt
  • Resolution: 0.1 ppm / 1 ppm / 0.01 ppt
  • Accuracy: 1 % Full Scale
  • TDS Calibration Points: Up to 3 Points (manual)
  • TDS Factor: 0.40 to 1.00
  • Salinity Range: 0.0 to 99.9 ppm, 100 to 999 ppm, 1.00 to 10.00 ppt, 0.0 to 1.00 %
  • Resolution: 0.1 ppm / 1 ppm / 0.01 ppt / 0.01%
  • Accuracy: 1 % Full Scale
  • Salinity Calibration Points: Up to 3 (manual)
  • Temperature Range: 0.0 to 50.0 °C / 32.0 to 120.0 °F
  • Resolution: 0.1 °C/°F
  • Accuracy: ±0.5°C/ ±0.9°F
  • Temperature Compensation: ATC /mTC
  • Non-Volatilememory: Yes
  • Auto-Off: 8.5mins after last key pressed (can be disabled)
  • LCD Display: Dual-display LCD (2.1 x 2.7cm)
  • Power: 4 X 1.5 V 'A76'micro alkaline batteries
  • Battery Life: > 150 hours
  • Dimensions; Weight: Boxed: 18.5 cm x 6.5 cm x 5 cm; 200 g
仕様
タイプMulti-Parameter Pocket Tester
Unit SizeEach