PIP2:PS Lipid Kinase Substrate
Thermo Scientific™

PIP2:PS Lipid Kinase Substrate

Specifically optimized for use with the Class I PI3 kinases (PIK3CA, PIK3CB, PIK3CD, and PIK3CG), this substrate consists of lipid vesicles of a 1:19 molar ratio of phosphoinositol-4,5-bisphosphate (PIP2) with phosphotidylserine (PS).
Have Questions?
Catalog NumberQuantity
PV5100400 nmol
Catalog number PV5100
Price (USD)
1,168.00
Each
Quantity:
400 nmol
Price (USD)
1,168.00
Each

Specifically optimized for use with the Class I PI3 kinases (PIK3CA, PIK3CB, PIK3CD, and PIK3CG), this substrate consists of lipid vesicles of a 1:19 molar ratio of phosphoinositol-4,5-bisphosphate (PIP2) with phosphotidylserine (PS).

For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Assay EntryBiochemical ADP accumulation
Gene ID (Entrez)5290, 5294
Quantity400 nmol
Shipping ConditionDry Ice
SubstratePhosphoinositol-4,5-bisphosphate (PIP2) lipid vesicles with phosphotidylserine (PS)
Target EntryPIK3CG, PIK3CD/PIK3R1, PIK3CA/PIK3R1
Validated KinasesPIK3CA, PIK3CG, PIK3CB, PIK3CD
Detection MethodFluorescence
Substrate PropertiesLipid-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeLipid Kinases
Unit SizeEach
Contents & Storage
PIP2:PS Lipid Kinase Substrate in 50 mM HEPES pH 7.5, 3 mM MgCl2, and 1 mM EGTA.

Store at -20°C.