Pre-Weigh Buffered Peptone Water (BPW) (ISO) Agar
実際の製品は異なる場合があります
Thermo Scientific™

Pre-Weigh Buffered Peptone Water (BPW) (ISO) Agar

Speed up your workflow when preparing medium for pre-enrichment of Salmonella species from foods with pre-weighed media sachets.
製品番号(カタログ番号)数量
PW1049-1L20 x Sachets for 1 L Medium
PW1049-8L10 x Sachets for 8 L Medium
製品番号(カタログ番号) PW1049-1L
価格(JPY)
-
数量:
20 x Sachets for 1 L Medium
Speed up your workflow when preparing microbial culture medium for pre-enrichment of Salmonella species from food samples with Thermo Scientific™ Pre-weigh Buffered Peptone Water (ISO) Agar. The pre-weighed sachets of Thermo Scientific™ Oxoid™ dehydrated culture media are designed for single use when preparing 1 L or 8 L volumes of your medium. Save valuable time and start testing quickly by removing the requirement to weigh powder—just rip, tip and pour.

Pre-Weigh Buffered Peptone Water (ISO) sachets contain the familiar Thermo Scientific Oxoid Buffered Peptone Water (ISO) medium (Manf. Part No. CM1049) and are part of a range of our most frequently used and well-characterized dehydrated culture media, which is now available in handy pre-weighed foil packages.

Simply tear open the selected media sachet, add to the required volume of water, mix, and autoclave as usual.

  • Efficient: choose from a full range of pre-weighed media formats for reduced hands-on prep time and maximum productivity
  • Convenient: rip and tip sachets enable faster media production and simplified workflows
  • Compliant: proven and validated media formats deliver confidence and reliable performance
  • Productive: reduce routine dehydrated culture media handling and weighing to spend more time on value-added tasks.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

実験室用。
仕様
概要Pre-Weigh Buffered Peptone Water (ISO)
数量20 x Sachets for 1 L Medium
形状粉末
製品タイプAgar
Unit SizeEach