Sodium Metasilicate Pentahydrate, Technical, Fisher Chemical™
Sodium Metasilicate Pentahydrate, Technical, Fisher Chemical™
Sodium Metasilicate Pentahydrate, Technical, Fisher Chemical™
Fisher Chemical™

Sodium Metasilicate Pentahydrate, Technical, Fisher Chemical™

Sodium Metasilicate Pentahydrate, Technical, Fisher Chemical, Quantity: 3kg, CAS: 10213-79-3, White, Melting Point: 72 deg.C, Molecular Formula: H10Na2O8Si, Molecular Weight: 212.137, Packaging: Plastic powder jar, pH: 12.6
製品番号(カタログ番号)数量
S-5360-63
または、製品番号S/5360/63
3 kg
製品番号(カタログ番号) S-5360-63
または、製品番号S/5360/63
価格(JPY)
-
数量:
3 kg
一括またはカスタム形式をリクエストする
仕様
CAS10213-79-3
色White
式量212.74g/mol
IUPAC名disodium;dioxido(oxo)silane;pentahydrate
InChIキーADPGKKZKGXANON-UHFFFAOYSA-N
さらに表示
Sodium Metasilicate Pentahydrate, Technical, Fisher Chemical, Quantity: 3kg, CAS: 10213-79-3, White, Melting Point: 72 deg.C, Molecular Formula: H10Na2O8Si, Molecular Weight: 212.137, Packaging: Plastic powder jar, pH: 12.6