Labtainer™ BioProcess Container (BPC), 100 L with 3 Ports, 2D BPC
Thermo Scientific™

Labtainer™ BioProcess Container (BPC), 100 L with 3 Ports, 2D BPC

The Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 100 L 2D pillow design is composed of CX5-14 single web, multi-layer plastic詳細を見る
製品番号(カタログ番号)数量
SH3066702100 L, each
製品番号(カタログ番号) SH3066702
価格(JPY)
-
数量:
100 L, each
The Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 100 L 2D pillow design is composed of CX5-14 single web, multi-layer plastic film and includes 3 panel ports.

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All Labtainers are constructed in an ISO 7 cleanroom under cGMP conditions.

Applications of the Thermo Scientific Labtainer BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors.
• Buffer, culture media, and process liquid preparation and storage.
• Chromatography feed and fraction collection.
• Harvest, storage and transport of bulk drug product and bulk drug precursors.

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library – with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and vent filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.

仕様
寸法Chamber: 71.12 cm x 76.2 cm (28 in. x 30 in.)
フィルム詳細CX5-14
使用対象 (装置)SV50076.03
材料Polyethylene
外部包装Packages placed in strong cardboard cartons with product and lot identification
ポート詳細Panel Ports: 2 x 0.38 in. 1 x 0.25 in.
製品ラインProductainer
保護用内部包装Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
数量100 L, each
滅菌方法Irradiated, Gamma Irradiated
配管ライン1Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
配管ライン2Line 2. MPC body, polycarbonate, Tubing: C-Flex, length: 61 cm (24 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.), Line 2. MPC insert, polycarbonate, Tubing: C-Flex, length: 30 cm (12 in.) ID x OD: 9.5 x 12.7mm (0.38 x 0.5 in.)
配管ライン3Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 66 cm (26 in.) ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
Unit Size100 L