Search
Search

| Catalog Number | Type |
|---|---|
| START-CP3 | Pint Bottle Cap |
| START-B00 | Burette Cover |
| START-B10 | 10 mL Burette |
| START-B20 | 20 mL Burette |
| START-B50 | 50 mL Burette |
| START-BT0 | Reagent Bottle Holder |
| START-BT1 | 1 L Plastic Bottle |
| START-BT2 | 1 L Glass Bottle |
| START-BT3 | 12 x 1 L Plastic Bottles |
| START-BT4 | 12 x 1 L Glass Bottles |
| START-CP1 | Reagent Bottle Cap (GL38) |
| START-CP2 | Reagent Bottle Cap (GL45) |
| START-DS1 | Dispenser Probe |
| START-DVK | Dispenser Verification Kit |
| START-EH1 | Electrode Holder Assembly |
| START-EH2 | Stopper |
| START-EH3 | Cable Management |
| START-PS1 | US/Japan Power Supply |
| START-PS2 | EU Power Supply |
| START-PS3 | UK/Singapore Power Supply |
| START-PS4 | ANZ Power Supply |
| START-PS5 | China Power Supply |
| START-TB1 | Standard Tubing Kit |
| START-TB2 | Light-blocking Tubing Kit |
| START-TB3 | Drying Tube |
| START-TB4 | Replacemet Burette |
| START-TB5 | Replacement Reagent Cap |
| START-TB6 | Replacement Dispenser Probe |
| START-UM1 | User Manual |
| START-USB | USB Cable |
| START-VK1 | Value Replacement |