TDE Series -40°C Ultra-Low Temperature Freezers - FAQs
TDE Series -40°C Ultra-Low Temperature Freezers - FAQs
View additional product information for TDE Series -40°C Ultra-Low Temperature Freezer - FAQs (TDE60040FV, TDE60040FA, TDE60040FD, TDE60040LD, TDE60040LV, TDE60040LA)