Detect urease activity in bacteria by observing a color change due to the hydrolysis of urea, which helps differentiate urease- producing Enterobacteriaceae from members of the Salmonella and Shigella groups with this medium.
Urea Broth (Christensen and Maslen) is used to detect urease activity in bacteria by observing a color change due to the hydrolysis of urea, which helps differentiate urease- producing Enterobacteriaceae from members of the Salmonella and Shigella groups with this medium.