Mitochondrial Membrane Potential Apoptosis Kit, with Mitotracker™ Red & Annexin V Alexa Fluor™ 488, for flow cytometry
Invitrogen™

Mitochondrial Membrane Potential Apoptosis Kit, with Mitotracker™ Red & Annexin V Alexa Fluor™ 488, for flow cytometry

This flow cytometry product provides a rapid and convenient assay for two hallmarks of apoptosis - phosphatidylserine externalization and changesRead more
Have Questions?
Catalog NumberQuantity
V351161 kit
Catalog number V35116
Price (USD)
537.65
Online exclusive
572.00
Save 34.35
1 kit
Quantity:
1 kit
Price (USD)
537.65
Online exclusive
572.00
Save 34.35
1 kit
This flow cytometry product provides a rapid and convenient assay for two hallmarks of apoptosis - phosphatidylserine externalization and changes in mitochondrial membrane potential. After staining a cell population with Alexa Fluor™ 488 annexin V and MitoTracker™ Red CMXRos dye in the provided binding buffer, live cells exhibit very little green fluorescence and bright red fluorescence, whereas apoptotic cells exhibit green fluorescence and decreased red fluorescence. These populations can easily be distinguished using a flow cytometer, and the 488 nm line of an argon-ion laser can be used to excite both dyes.

View a selection guide for all apoptosis assays for flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Excitation/EmissionMitotracker™ red 579⁄599, Alexa Fluor™ 488: 499⁄521
Flow Cytometer Laser Lines488
For Use With (Application)Flow Cytometry
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
No. of Reactions50
Product LineAlexa Fluor, MitoTracker
Product TypeMitochondrial Membrane Apoptosis Kit
Quantity1 kit
Shipping ConditionWet Ice
ConjugateAlexa Fluor 488, MitoTracker Red
FormatTube, Slide
Unit Size1 kit
Contents & Storage
Contains 1 vial of annexin, Alexa Fluor™ 488 conjugate (250 μL), 3 vials of MitoTracker™ red (50 μg per vial), 1 bottle of annexin binding buffer (5X solution, 15 mL), and 1 vial of DMSO (100 μL).

Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

What are the fluorescence excitation/emission maxima for the dyes contained in the Mitochondrial Membrane Potential Apoptosis Kit, with Mitotracker Red & Alexa Fluor 488 annexin V, for flow cytometry

Alexa Fluor 488 annexin V: 499/521 nm
MitoTracker Red: 579/599 nm

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to study apoptosis using an Annexin V conjugate, but with adherent cells via microscopy instead of flow cytometry. Can this be done?

It has been done, but we don‘t recommend it. Both healthy cells and apoptotic cells possess phosphatidylserine on the cell surface, which can be detected with Annexin V, but apoptotic cells have significantly more of it. You can easily tell the difference between these two populations with flow cytometry, because flow cytometers are more sensitive and have a higher throughput. But with a microscope, you cannot always tell the difference, especially for adherent cells. Instead, for microscopy, we recommend a different technique, such as detecting caspases with CellEvent Caspase Detection Reagents.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (4)

Citations & References
Abstract
Galectin 1 modulates plasma cell homeostasis and regulates the humoral immune response.
Authors:Anginot A, Espeli M, Chasson L, Mancini SJ, Schiff C,
Journal:J Immunol
PubMed ID:23616571
'Galectin-1 (GAL1) is an S-type lectin with multiple functions, including the control of B cell homeostasis. GAL1 expression was reported to be under the control of the plasma cell master regulator BLIMP-1. GAL1 was detected at the protein level in LPS-stimulated B cells and was shown to promote Ig secretion ... More
FGF23 activates injury-primed renal fibroblasts via FGFR4-dependent signalling and enhancement of TGF-ß autoinduction.
Authors:Smith ER, Holt SG, Hewitson TD
Journal:Int J Biochem Cell Biol
PubMed ID:28919046
Bone-derived fibroblast growth factor 23 (FGF23) is an important endocrine regulator of mineral homeostasis with effects transduced by cognate FGF receptor (FGFR)1-a-Klotho complexes. Circulating FGF23 levels rise precipitously in patients with kidney disease and portend worse renal and cardiovascular outcomes. De novo expression of FGF23 has been found in the ... More
High Tidal Volume Induces Mitochondria Damage and Releases Mitochondrial DNA to Aggravate the Ventilator-Induced Lung Injury.
Authors:Lin JY, Jing R, Lin F, Ge WY, Dai HJ, Pan L
Journal:Front Immunol
PubMed ID:30018615
This study aimed to determine whether high tidal volume (HTV) induce mitochondria damage and mitophagy, contributing to the release of mitochondrial DNA (mtDNA). Another aim of the present study was to investigate the role and mechanism of mtDNA in ventilator-induced lung injury (VILI) in rats. ... More
Adaptive phenotypic modulations lead to therapy resistance in chronic myeloid leukemia cells.
Authors:Baykal-Köse S, Acikgoz E, Yavuz AS, Gönül Geyik Ö, Ates H, Sezerman OU, Özsan GH, Yüce Z
Journal:PLoS One
PubMed ID:32106243
Tyrosine kinase inhibitor (TKI) resistance is a major problem in chronic myeloid leukemia (CML). We generated a TKI-resistant K562 sub-population, K562-IR, under selective imatinib-mesylate pressure. K562-IR cells are CD34-/CD38-, BCR-Abl-independent, proliferate slowly, highly adherent and form intact tumor spheroids. Loss of CD45 and other hematopoietic markers reveal these cells have ... More