Europium ingot, 99.9% (REO)
Europium ingot, 99.9% (REO)
Europium ingot, 99.9% (REO)
Thermo Scientific Chemicals

Europium ingot, 99.9% (REO)

CAS: 7440-53-1 | Eu | 151.96 g/mol
製品番号(カタログ番号) 000120.06
または、製品番号000120-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-53-1
IUPAC Nameeuropium
Molecular FormulaEu
InChI KeyOGPBJKLSAFTDLK-UHFFFAOYSA-N
SMILES[Eu]
さらに表示
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under mineral oil
RUO – Research Use Only