Iodine, crystalline, 99.99+% (metals basis)
Iodine, crystalline, 99.99+% (metals basis)
Iodine, crystalline, 99.99+% (metals basis)
Thermo Scientific Chemicals

Iodine, crystalline, 99.99+% (metals basis)

CAS: 7553-56-2 | I2 | 253.81 g/mol
製品番号(カタログ番号) 000158.18
または、製品番号000158-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
さらに表示
FormCrystals
Non-Volatile Residue≤0.005%
Impurity contentChlorine and Bromine (as Cl): ≤0.005%
Appearance (Color)Dark grey to black to purple
Assay from Supplier's CofA≥99.99% Iodine (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only