Lanthanum pieces, 99.9% (REO)
Lanthanum pieces, 99.9% (REO)
Lanthanum pieces, 99.9% (REO)
Thermo Scientific Chemicals

Lanthanum pieces, 99.9% (REO)

CAS: 7439-91-0 | La | 138.91 g/mol
製品番号(カタログ番号) 000175.14
または、製品番号000175-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7439-91-0
IUPAC Namelanthanum
Molecular FormulaLa
InChI KeyFZLIPJUXYLNCLC-UHFFFAOYSA-N
SMILES[La]
さらに表示
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only