Terbium pieces, 99.9% (REO)
Terbium pieces, 99.9% (REO)
Terbium pieces, 99.9% (REO)
Thermo Scientific Chemicals

Terbium pieces, 99.9% (REO)

CAS: 7440-27-9 | Tb | 158.93 g/mol
製品番号(カタログ番号) 000324.03
または、製品番号000324-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-27-9
IUPAC Nameterbium
Molecular FormulaTb
InChI KeyGZCRRIHWUXGPOV-UHFFFAOYSA-N
SMILES[Tb]
さらに表示
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only