Zinc wire, 0.25mm (0.01in) dia, 99.99+% (metals basis)
Zinc wire, 0.25mm (0.01in) dia, 99.99+% (metals basis)
Zinc wire, 0.25mm (0.01in) dia, 99.99+% (metals basis)
Thermo Scientific Chemicals

Zinc wire, 0.25mm (0.01in) dia, 99.99+% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol
製品番号(カタログ番号) 000648.G1
または、製品番号000648-G1
価格(JPY)
-
数量:
1 m
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
さらに表示
FormWire, 0.25 mm diameter
Total Metal Impurities<0.01%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only