Zinc sulfate hydrate, Puratronic, 99.998% (metals basis)
Zinc sulfate hydrate, Puratronic, 99.998% (metals basis)
Zinc sulfate hydrate, Puratronic, 99.998% (metals basis)
Thermo Scientific Chemicals

Zinc sulfate hydrate, Puratronic, 99.998% (metals basis)

CAS: 16788-42-4 | H2O5SZn | 179.45 g/mol
製品番号(カタログ番号) 011138.09
または、製品番号011138-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16788-42-4
IUPAC Namezinc(2+) hydrate sulfate
Molecular FormulaH2O5SZn
InChI KeyRNZCSKGULNFAMC-UHFFFAOYSA-L
SMILESO.[Zn++].[O-]S([O-])(=O)=O
さらに表示
FormPowder or crystals or crystalline powder
Total Metal Impurities≤0.002%
Zinc sulfate hydrate is used in the production of rayon, as a feed supplement, and as a fertilizer ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only