Silver sulfide, 99%
Silver sulfide, 99%
Silver sulfide, 99%
Thermo Scientific Chemicals

Silver sulfide, 99%

CAS: 21548-73-2 | Ag2S | 247.80 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 011416.06
または、製品番号011416-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS21548-73-2
IUPAC Namedisilver(1+) sulfanediide
Molecular FormulaAg2S
InChI KeyXUARKZBEFFVFRG-UHFFFAOYSA-N
SMILES[S--].[Ag+].[Ag+]
さらに表示
Appearance (Color)Dark grey to black
FormPowder
Assay from Suppliers CofA≥98.5%
Silver sulfide is useful as a photosensitizer in photography, Intermediates, chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only