Silver wire, 0.5mm (0.02in) dia, 1/2 hard, 99.9% (metals basis)
Silver wire, 0.5mm (0.02in) dia, 1/2 hard, 99.9% (metals basis)
Silver wire, 0.5mm (0.02in) dia, 1/2 hard, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver wire, 0.5mm (0.02in) dia, 1/2 hard, 99.9% (metals basis)

Form: Approx. 2.06g/m, Temper: 1/2 hard | CAS: 7440-22-4 | Ag | 107.87 g/mol
製品番号(カタログ番号) 011433.G6
または、製品番号011433-G6
価格(JPY)
-
数量:
10 m
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
さらに表示
Appearance (Color)Silver
FormWire, 0.5mm (0.02in) diameter, 1/2 hardened.
Assay from Supplier's CofA>99.9% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only