Molybdenum silicide, Mo 61% min, typically 99.5% (metals basis)
Molybdenum silicide, Mo 61% min, typically 99.5% (metals basis)
Molybdenum silicide, Mo 61% min, typically 99.5% (metals basis)
Thermo Scientific Chemicals

Molybdenum silicide, Mo 61% min, typically 99.5% (metals basis)

CAS: 12136-78-6 | MoSi2 | 152.12 g/mol
製品番号(カタログ番号) 011549.18
または、製品番号011549-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12136-78-6
IUPAC Name(silanylidenemolybdenio)silane
Molecular FormulaH4MoSi2
InChI KeyREHMQSAMQCFKDZ-UHFFFAOYSA-N
SMILES[SiH2]=[Mo]=[SiH2]
さらに表示
Assay (unspecified)Mo 61% min
In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehiclesMolybdenum silicide is used in electric furnaces, glass, steel, electronics and ceramics industries. It finds application in heat treatment materials. It is also used in microelectronics as a contact material. Further, it serves as a dry lubricant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehicles
RUO – Research Use Only