Thermo Scientific Chemicals

Potassium fluoride dihydrate, 98.5+%, Thermo Scientific Chemicals

製品番号(カタログ番号): 011555.14
25 g, Each
Thermo Scientific Chemicals

Potassium fluoride dihydrate, 98.5+%, Thermo Scientific Chemicals

製品番号(カタログ番号): 011555.14
25 g, Each
数量
製品番号(カタログ番号): 011555.14

化学物質識別子

CAS
13455-21-5
IUPAC Name
potassium dihydrate fluoride
Molecular Formula
FH4KO2
InChI Key
MZFXHSQFQLKKFD-UHFFFAOYSA-M
SMILES
O.O.[F-].[K+]
Assay (unspecified)
98.6% min

概要

Potassium fluoride dihydrate is used as a stationary phase in gas chromatography. It is also used for the conversion of chlorocarbons into fluorocarbons through Finkelstein reaction. Further, it is used as a fluxing agent of silver brazing and metallurgical processing. It serves as an analytical reagent, complexing agent and masking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text