Silver foil, 0.025mm (0.001in) thick, annealed, 99.95% (metals basis)
Silver foil, 0.025mm (0.001in) thick, annealed, 99.95% (metals basis)
Silver foil, 0.025mm (0.001in) thick, annealed, 99.95% (metals basis)
Thermo Scientific Chemicals

Silver foil, 0.025mm (0.001in) thick, annealed, 99.95% (metals basis)

Form: Approx. 5.9g/150 x 150mm; Temper: soft | CAS: 7440-22-4 | Ag | 107.87 g/mol
数量:
150 x 150 mm
150 x 600 mm
150 x 1000 mm
製品番号(カタログ番号) 012190.GT
または、製品番号012190-GT
価格(JPY)
-
数量:
150 x 150 mm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
さらに表示
Appearance (Color)Silver
Total Metal Impurities≤ 0.05%
Thickness0.022-0.028mm
FormFoil
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only