Indium(III) oxide, 99.9% (metals basis)
Indium(III) oxide, 99.9% (metals basis)
Indium(III) oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Indium(III) oxide, 99.9% (metals basis)

CAS: 1312-43-2 | In2O3 | 277.63 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) 012581.30
または、製品番号012581-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1312-43-2
IUPAC Namediindium(3+) trioxidandiide
Molecular FormulaIn2O3
InChI KeyPJXISJQVUVHSOJ-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[In+3].[In+3]
さらに表示
Total Metal Impurities0.1% max.
Indium(III) oxide is widely utilized as a n-type semiconductor, which is used in integrated circuits as a resistive element. It finds applications in batteries and as a part of some stain formulations. It is used as thin film coatings in optical, antistatic and infrared reflectors. It is doped with tin oxide and used as transparent conductive coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only