Hexacarbonylchromium, 99%
Hexacarbonylchromium, 99%
Hexacarbonylchromium, 99%
Thermo Scientific Chemicals

Hexacarbonylchromium, 99%

CAS: 13007-92-6 | C6CrO6 | 220.056 g/mol
製品番号(カタログ番号) 013061.18
または、製品番号013061-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13007-92-6
IUPAC Namehexakis(methanidylidyneoxidanium) chromium
Molecular FormulaC6CrO6
InChI KeyKOTQLLUQLXWWDK-UHFFFAOYSA-N
SMILES[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
さらに表示
Assay (unspecified)98.6% min
Hexacarbonylchromium is widely used as a catalyst for isomerization of alkenes. It is involved in the hydrogenation of 1,3-conjugated dienes and oxidation of allylic and benzylic positions. It acts as a precursor to prepare phenanthrenes, tricarbonyl(arene)chromium and Fischer carbine complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only