Praseodymium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Praseodymium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Praseodymium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Praseodymium(III) sulfate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

製品番号(カタログ番号)数量
013180.14
または、製品番号013180-14
25 g
製品番号(カタログ番号) 013180.14
または、製品番号013180-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13510-41-3
IUPAC Namedipraseodymium(3+) octahydrate trisulfate
Molecular FormulaH16O20Pr2S3
InChI KeyNIJXLQIGVGZNKP-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.[Pr+3].[Pr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
さらに表示
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only