Di-n-butyltin dichloride, 96%
Di-n-butyltin dichloride, 96%
Di-n-butyltin dichloride, 96%
Thermo Scientific Chemicals

Di-n-butyltin dichloride, 96%

Organotin intermediate | CAS: 683-18-1 | C8H18Cl2Sn | 303.842 g/mol
数量:
10 g
250 g
1 kg
製品番号(カタログ番号) 014116.A1
または、製品番号014116-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS683-18-1
Molecular FormulaC8H20Cl2Sn
InChI KeyGSCBVGBIRKBMAC-UHFFFAOYSA-N
SMILESCl.Cl.CCCC[SnH2]CCCC
Molecular Weight (g/mol)305.86
Assay (unspecified)>95.0%
Organotin intermediate.Di-n-butyltin dichloride is used as an organotin intermediate. It serves as heat and light stabilizer for polyvinyl chloride plastics. It is also used in the preparation of methyl 2-(methoxycarbonylmethyl)-5-iodobenzene by reacting with 2,5-diiodobenzoic acid. It acts as an esterification catalyst and a veterinary vermicide and tapeworm remedy. Further, it is used as ion exchange agent, process regulators and processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only