Tin(II) 2-ethylhexanoate, tech.
Tin(II) 2-ethylhexanoate, tech.
Tin(II) 2-ethylhexanoate, tech.
Thermo Scientific Chemicals

Tin(II) 2-ethylhexanoate, tech.

CAS: 301-10-0 | C16H30O4Sn | 405.122 g/mol
数量:
100 g
500 g
2 kg
製品番号(カタログ番号) 018590.22
または、製品番号018590-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS301-10-0
IUPAC Nameλ²-tin(2+) bis(2-ethylhexanoate)
Molecular FormulaC16H30O4Sn
InChI KeyKSBAEPSJVUENNK-UHFFFAOYNA-L
SMILES[Sn++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
さらに表示
Appearance (Color)Pale yellow
Refractive Index1.4920 to 1.4960
Formliquid
Assay (Titration)Total Tin: 28.0 wt%, min
Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, lubricants, addition agents and stabilizers for transformer oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only