Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals
Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals
Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals

製品番号(カタログ番号) 018859.18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS18267-08-8
IUPAC Namezirconium(4+) ethanolate
Molecular FormulaC2H5OZr
InChI KeyVCYRFVPNTBNCLW-UHFFFAOYSA-N
SMILES[Zr+4].CC[O-]
さらに表示
Appearance (Color)White to pale cream
FormCrystalline powder or powder and/or chunks
Assay from Suppliers CofA≥98.5%
Zirconium(IV) ethoxide is used as precursor for the synthesis of olefin-functionalized zirconium-containing monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only