n-Pentane, HPLC Grade, 99% min
n-Pentane, HPLC Grade, 99% min
n-Pentane, HPLC Grade, 99% min
Thermo Scientific Chemicals

n-Pentane, HPLC Grade, 99% min

Maximum level of impurities: Evaporation residue 3ppm, H2O 0.01% | CAS: 109-66-0 | C5H12 | 72.15 g/mol
数量:
1 L
4 L
4 x 1 L
4 x 4 L
製品番号(カタログ番号) 022907.K7
または、製品番号022907-K7
価格(JPY)
-
数量:
4 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
さらに表示
CommentSub micron filtered
Optical Absorbance at210nm: ≤0.5 AU
Assay (GC)≥99.9% ( As C5 Hydrocarbons)
Optical Absorbance at200nm: ≤1.0 AU
Optical Absorbance at245nm: ≤0.01 AU
さらに表示
n-Pentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only