N,N-Dimethylformamide, HPLC Grade, 99.7+%
N,N-Dimethylformamide, HPLC Grade, 99.7+%
N,N-Dimethylformamide, HPLC Grade, 99.7+%
Thermo Scientific Chemicals

N,N-Dimethylformamide, HPLC Grade, 99.7+%

CAS: 68-12-2 | C3H7NO | 73.10 g/mol
数量:
4 L
4 x 1 L
4 x 4 L
6 x 1 L
製品番号(カタログ番号) 022915.K7
または、製品番号022915-K7
価格(JPY)
-
数量:
4 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
さらに表示
Residual water0.05%
Residue on Evaporation5ppm
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films. It is used as a mobile phase in High Performance Liquid Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only