Dichloromethane, HPLC Grade, 99.7+%, stab. with amylene
Dichloromethane, HPLC Grade, 99.7+%, stab. with amylene
Dichloromethane, HPLC Grade, 99.7+%, stab. with amylene
Thermo Scientific Chemicals

Dichloromethane, HPLC Grade, 99.7+%, stab. with amylene

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
数量:
250 mL
1 L
4 L
4 x 1 L
製品番号(カタログ番号) 022917.K7
または、製品番号022917-K7
価格(JPY)
-
数量:
4 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
さらに表示
Optical Absorbance at235nm: 0.55AU, max
FormLiquid
Impurity content5ppt, max as Heptachlor epoxide by GC-ECD
Impurity contentFree halogens P.T.
Optical Absorbance at240nm: 0.20AU, max
さらに表示
Dichloromethane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only